EMAIL THIS PAGE TO A FRIEND
International journal of phytoremediation
Effects of the halophytes Tecticornia indica and Suaeda fruticosa on soil enzyme activities in a Mediterranean Sabkha.
PMID 23487995
Abstract
In the present work, we studied the effectiveness of the predominant halophytes of Soliman sabkha (Tecticornia indica and Suaeda fruticosa) to promote soil biological activities and ecosystem productivity. Soil Arylsulphatese ARY, beta-glucosidase beta-GLU, phosphatase PHO, invertase INV, urease URE, and dehydogenase DES activities in Extra- and Intra-tuft halophytes and plant productivity were assessed. Results revealed a high increase of microbial community and ARY, beta-GLU, PHO, INV URE and DES activities (+298%, +400%, +800%, +350%, +320%, +25% and +759%, respectively) in Intra-tuft rhizosphere as compared to Extra-tuft one, which is likely due to the significant decrease of salinity in the rhizosphere of Tecticornia indica and Suaeda fruticosa. Both perennial plants exhibited high productivities (7.4 t dry weight ha(-1) and 2.2 t dry weight ha(-1), respectively) and Na+-hyperaccumulating capacities (0.75 t Na+ ha(-1) and 0.22 t Na+ ha(-1), respectively), reducing salt constraint and favouring soil fertility. This constitutes a promising alternative to enhance productivity in such a salt-affected biotope by offering suitable microhabitat for annual glycophytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I4504 Invertase from baker's yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
I9274 Invertase from baker's yeast (S. cerevisiae), 200-300 units/mg solid
I4753
Invertase from Candida utilis, Grade X, ≥300 units/mg solid
I0408 Invertase Glycoprotein Standard, BioReagent, from Saccharomyces cerevisiae, for proteomics
S9754 Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid
S1629 Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
S9626 Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
S9751 Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
S8629 Sulfatase from Patella vulgata (keyhole limpet), Type V, essentially salt-free, lyophilized powder, ≥5 units/mg solid
S8504 Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
U1500 Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
U4002 Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
U0251 Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
U1875 Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
U7752 Urease from Canavalia ensiformis (Jack bean)