EMAIL THIS PAGE TO A FRIEND
International journal of phytoremediation
Effects of Cd, Pb, Zn, Cu-resistant endophytic Enterobacter sr CBSB1 and Rhodotorula sp. CBSB79 on the growth and phytoextraction of Brassica plants in multimetal contaminated soils.
PMID 23488174
Abstract
To survey the effects of endophytic Enterobacter sp. CBSB1 and Rhodotorula sp. CBSB79 resistant to Cd2+, Pb2+, Zn2+, and Cu2+ on the growth and phytoextraction of Brassica, the endophytes were isolated by surface- sterilized methods and characterized. The CBSB1 significantly increased 44.2% of the dry weight of Brassica napus in the multimetal contaminated soil (P < 0.05) and showed no effect or declined the dry weight of B. alboglabra, B. campestris ssp. chinensis var. cummunis, B. campestris ssp. chinensis var. utilis cv. Youqing12, B. campestris ssp. chinensis var. utilis cv. Lvbao701 plants. The dry weights of B. napus, B. campestris ssp. chinensis var. utilis, and B. alboglabra showed a significant increase when the CBSB79 was inoculated (P < 0.05). In general, inoculation with bacteria and yeast did not greatly alter the metal concentration in plant tissues. Compared to Enterobacter sp. CBSB1, the yeast Rhodotorula sp CBSB79 showed higher potentials to improve extraction efficacy of Cd, Pb, Zn, and Cu by Brassica seedlings in the field.