EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel metastable metallic and semiconducting germaniums.
PMID 23492980
Abstract
Group-IVa elements silicon and germanium are known for their semiconducting properties at room temperature, which are technologically critical. Metallicity and superconductivity are found at higher pressures only, Ge β-tin (tI4) being the first high-pressure metallic phase in the phase diagram. However, recent experiments suggest that metallicity in germanium is compatible with room conditions, calling for a rethinking of our understanding of its phase diagram. Missing structures can efficiently be identified based on structure prediction methods. By means of ab initio metadynamics runs we explored the lower-pressure region of the phase diagram of germanium. A monoclinic germanium phase (mC16) with four-membered rings, less dense than diamond and compressible into β-tin phase (tI4) was found. Tetragonal bct-5 appeared between diamond and tI4. mC16 is a narrow-gap semiconductor, while bct-5 is metallic and potentially still superconducting in the very low pressure range. This finding may help resolving outstanding experimental issues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
636428
Diamond, nanopowder, <10 nm particle size (TEM), ≥97% trace metals basis
C
636444
Diamond, nanopowder, <10 nm particle size (TEM), ≥95% trace metals basis
C
483575 Diamond, monocrystalline powder, ~1 μm
C
483591 Diamond, synthetic monocrystalline powder, ≤1 μm
C
483583 Diamond, synthetic polycrystalline powder, ~1 μm
C
07745 Diamond nanoparticles suspension, ~1% (w/w) solid material basis
327395
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Ge
203351
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Ge
203343
Germanium, chips, 99.999% trace metals basis
Ge
263230
Germanium, chips, 99.999% trace metals basis
Ge
14509
Tin, ≥99%, powder
Sn
265640
Tin, powder, <150 μm, 99.5% trace metals basis
Sn
576883
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sn
243434
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sn
204692
Tin, shot, 99.999% trace metals basis
Sn
265632
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sn
217697
Tin, 99.8%, shot, 3 mm
Sn
520373
Tin, powder, 10 μm, 99% trace metals basis
Sn
755702
Tin, powder, -100 mesh, 99.99% trace metals basis
Sn
265756
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sn
356956
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sn
356948
Tin, foil, thickness 0.127 mm, 99.9%
Sn
67226
Tin (impurities), manufactured by MBH Analytical Ltd (71X SR0)
Sn