EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
Autocrine amplification of integrin αIIbβ3 activation and platelet adhesive responses by deoxyribose-1-phosphate.
PMID 23494007
Abstract
Using direct injection mass spectrometry (DIMS) we discovered that deoxyribose-1-phosphate (dRP) is released by platelets upon activation. Interestingly, the addition of exogenous dRP to human platelets significantly increased platelet aggregation and integrin αIIbβ3 activation in response to thrombin. In parallel, genetically modified platelets with double genetic deletion of thymidine phosphorylase and uridine phosphorylase were characterised by reduced release of dRP, impaired aggregation and decreased integrin αIIbβ3 activation in response to thrombin. In vitro platelet adhesion onto fibrinogen and collagen under physiological flow conditions was potentiated by treatment of human platelets with exogenous dRP and impaired in transgenic platelets with reduced dRP release. Human and mouse platelets responded to dRP treatment with a sizeable increase in reactive oxygen species (ROS) generation and the pre-treament with the antioxidant apocynin abolished the effect of dRP on aggregation and integrin activation. Experiments directly assessing the activation of the small G protein Rap1b and protein kinase C suggested that dRP increases the basal levels of activity of these two pivotal platelet-activating pathways in a redox-dependent manner. Taken together, we present evidence that dRP is a novel autocrine amplifier of platelet activity, which acts on platelet redox levels and modulates integrin αIIbβ3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6539
2-Deoxy-α-D-ribose 1-phosphate bis(cyclohexylammonium) salt, ≥97%
C5H11O7P · 2C6H13N