EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Scanning electron microscopy evaluation of endothelialized tissue-engineered constructs.
PMID 23494441
Abstract
Scanning electron microscopy (SEM) is an important technique for evaluation of the efficiency of endothelialization of tissue-engineered constructs incorporating a surface endothelial cell layer. Here, we describe methodologies for the preparation of such constructs for SEM analysis that are applicable to a broad range of tissue-engineered constructs.