EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Formation of molybdate ion clusters by phosphomolybdic anions under matrix-assisted laser desorption/ionization conditions in the gas phase.
PMID 23494791