EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Effect of phosphoric acid as a matrix additive in matrix-assisted laser desorption/ionization analysis.
PMID 23495031
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5129
Gentisic acid sodium salt hydrate, ≥98%
C7H5NaO4