EMAIL THIS PAGE TO A FRIEND
Macromolecular Rapid Communications
Photoluminescent toroids formed by temperature-driven self-assembly of rhodamine B end-capped poly(N-isopropylacrylamide).
PMID 23495104