EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Evaluation of transport parameters for PVC based polyvinyl alcohol Ce(IV) phosphate composite membrane.
PMID 23498270
Abstract
The aim of this study was to investigate the preparation of novel membrane and the characterization of their properties. A new class of polyvinyl chloride (PVC) based polyvinyl alcohol Ce(IV) phosphate composite membrane was successfully prepared by solution casting method. The structural formation was confirmed by X-ray diffraction (XRD), Fourier transform infrared (FTIR) spectroscopy and morphological studies. The thermal property was investigated by thermogravimetry analysis (TGA) method. The order of surface charge density for various electrolytes was found to be LiCl