EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
Dermatic Scedosporium apiospermum infection after autologous bone marrow transplantation.
PMID 23503412
Abstract
Infection with Scedosporium apiospermum (S. apiospermum) is rare, although it is associated with a high fatality rate, especially in immunosuppressed patients. A 23-year-old man with acute myelogenous leukemia (AML) (M2) who was pretreated with chemotherapy for autologous bone marrow transplantation developed a skin ulcer on the left groin. After a culture study demonstrated the presence of S. apiospermum, voriconazole was administered and the lesion rapidly improved. Since a diagnosis of S. apiospermum continues to depend on the results of a fungal culture and most isolates of S. apiospermum are resistant to amphotericin B, voriconazole should be considered as the first choice when "mold" is thought to be the causative organism.