EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Separation of 20(S)-ginsenoside-Rg2 and its 20(R) epimer by reversed phase low pressure column (Rp-18) chromatography].
PMID 2350426
Abstract
20(S)-Ginsenoside-Rg2 and its 20(R) epimer were successfully separated using reversed-phase low pressure column (Rp-18) chromatography. The method has proved very convenient and rapid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08171
Ginsenoside Rg2, analytical standard
C42H72O13