EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Designed self-assembling peptides as templates for the synthesis of metal nanoparticles.
PMID 23504425
Abstract
Self-assembling peptides are water soluble and form biocompatible nanostructures under mild conditions through non-covalent interactions. They form supramolecular structures such as ribbons, nanotubes, and fibrils. Of particular interest is the possibility of using these peptide fibrils as templates for the growth of inorganic materials, such as metallic nanoparticles. The ability to reliably produce metal-coated fibrils with robust binding of metal nanoparticles is a vital first step towards the exploitation of these fibrils as conducting nanowires with applications in nano-circuitry. One promising strategy consists of the rational introduction of metal-binding amino acids (such as cysteine) at the level of the peptide building block. Upon assembly of the building blocks into fibrils, cysteine residues that remain accessible at the outside of the fibril core could serve as nucleation sites for metals. We will review in this chapter a case study of rationally designed cysteine-containing peptides and basic protocols for their metallization with silver, gold, and platinum nanoparticles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
481130
Gold(I) chloride, 99.9% trace metals basis
AuCl
379948
Gold(III) chloride, ≥99.99% trace metals basis
AuCl3
334049
Gold(III) chloride, 99%
AuCl3
373214
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Pt
373222
Platinum, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Pt
373230
Platinum, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Pt
349372
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Pt
267260
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Pt
305189
Platinum, foil, thickness 0.5 mm, 99.9% trace metals basis
Pt
349321
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Pt
327425
Platinum, foil, thickness 0.25 mm, 99.9% trace metals basis
Pt
349348
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Pt
327433
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Pt
267252
Platinum, foil, thickness 0.1 mm, 99.99% trace metals basis
Pt
349380
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Pt
349356
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Pt
303798
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Pt
349364
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Pt
267244
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Pt
298107
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Pt
298093
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Pt
685453
Platinum, nanopowder, <50 nm particle size (TEM)
Pt
204048
Platinum, powder (coarse), 99.99% trace metals basis
Pt
204013
Platinum, powder, 99.995% trace metals basis
Pt
327476
Platinum, powder, ≤10 μm, 99.9% trace metals basis
Pt
327484
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Pt
267155
Platinum, sponge, ≥99.9% trace metals basis
Pt
444685
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Pt
349399
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Pt
267236
Platinum, wire, diam. 1.0 mm, 99.99% trace metals basis
Pt