EMAIL THIS PAGE TO A FRIEND
Journal of cutaneous pathology
Granulomatous alopecia: a variant of alopecia areata?
PMID 23506008
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8895
Clofazimine
C27H22Cl2N4