EMAIL THIS PAGE TO A FRIEND
Molecular aspects of medicine
SLC17: a functionally diverse family of organic anion transporters.
PMID 23506876
Abstract
Molecular studies have determined that the SLC17 transporters, a family of nine proteins initially implicated in phosphate transport, mediate the transport of organic anions. While their role in phosphate transport remains uncertain, it is now clear that the transport of organic anions facilitated by this family of proteins is involved in diverse processes ranging from the vesicular storage of the neurotransmitters, to urate metabolism, to the degradation and metabolism of glycoproteins.