EMAIL THIS PAGE TO A FRIEND
Journal of cardiothoracic and vascular anesthesia
Pro: cerebral oximetry should be a routine monitor during cardiac surgery.
PMID 23507018
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295698
Sulfur dioxide, ≥99.9%
O2S