EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A silver complex of N,N'-disubstituted cyclic thiourea as an anti-inflammatory inhibitor of IκB kinase.
PMID 23508073
Abstract
A silver complex of N,N'-disubstituted cyclic thiourea inhibits inflammatory cytokine-stimulated NF-κB activity via IκB kinase inactivation.