EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
In situ continuous growth formation of synthetic biominerals.
PMID 23508225
Abstract
Continuous self-assembled growth of both the organic and inorganic components of materials with nacre-like structure is achieved upon mineralisation of chitin and chitosan scaffolds using a combined soaking method and the inclusion of poly(acrylic acid) and chitosan oligomers as additives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9213
Chitin from shrimp shells, practical grade, coarse flakes
C7170
Chitin from shrimp shells, practical grade, powder
C9752
Chitin from shrimp shells, BioReagent, suitable for analysis of chitinase, purified powder
81117
Poly(acrylic acid sodium salt), analytical standard, for GPC, 1,250
81123
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
81131
Poly(acrylic acid sodium salt), analytical standard, for GPC, 83,000
420344
Poly(acrylic acid sodium salt), average Mw ~2,100
447013
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
181293
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
323667
Poly(acrylic acid)
181285
Poly(acrylic acid), average Mv ~450,000
306215
Poly(acrylic acid), average Mv ~1,250,000
306223
Poly(acrylic acid), average Mv ~3,000,000
306231
Poly(acrylic acid), average Mv ~4,000,000
535931
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
523925
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
416002
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
416010
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
416029
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
416037
Poly(acrylic acid, sodium salt) solution, average Mw ~15,000, 35 wt. % in H2O