EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Double-strand break repair by homologous recombination in primary mouse somatic cells requires BRCA1 but not the ATM kinase.
PMID 23509290
Abstract
Homology-directed repair (HDR) is a critical pathway for the repair of DNA double-strand breaks (DSBs) in mammalian cells. Efficient HDR is thought to be crucial for maintenance of genomic integrity during organismal development and tumor suppression. However, most mammalian HDR studies have focused on transformed and immortalized cell lines. We report here the generation of a Direct Repeat (DR)-GFP reporter-based mouse model to study HDR in primary cell types derived from diverse lineages. Embryonic and adult fibroblasts from these mice as well as cells derived from mammary epithelium, ovary, and neonatal brain were observed to undergo HDR at I-SceI endonuclease-induced DSBs at similar frequencies. When the DR-GFP reporter was crossed into mice carrying a hypomorphic mutation in the breast cancer susceptibility gene Brca1, a significant reduction in HDR was detected, showing that BRCA1 is critical for HDR in somatic cell types. Consistent with an HDR defect, Brca1 mutant mice are highly sensitive to the cross-linking agent mitomycin C. By contrast, loss of the DSB signaling ataxia telangiectasia-mutated (ATM) kinase did not significantly alter HDR levels, indicating that ATM is dispensable for HDR. Notably, chemical inhibition of ATM interfered with HDR. The DR-GFP mouse provides a powerful tool for dissecting the genetic requirements of HDR in a diverse array of somatic cell types in a normal, nontransformed cellular milieu.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6142 Acc I from Acinetobacter calcoaceticus, Restriction Enzyme
R6885 Alu I from Arthrobacter luteus, Restriction Enzyme
R8631
Bcl I from Bacillus caldolyticus, Restriction Enzyme
R6377
Bgl II from Bacillus licheniformis, Restriction Enzyme
R3131
Bln I from Brevibacterium linens, buffered aqueous glycerol solution
R3635
Bsm I from Bacillus stearothermophilus NUB 36, Restriction Enzyme
R4253 BstE II from Bacillus stearothermophilus ET, Restriction Enzyme
R1761
Cfo I from Clostridium formicoaceticum, Restriction Enzyme
R7763
Cla I from Caryophanon latum L, Restriction Enzyme
R4256
Dde I from Desulfovibrio desulfuricans strain Norway, Restriction Enzyme
R4381
Dra I from Deinococcus radiophilus, Restriction Enzyme
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme
R6265
EcoR I from Escherichia coli BS5, Restriction Enzyme
R2756
EcoR V from Escherichia coli, buffered aqueous glycerol solution
R5628 Hae III from Haemophilus aegyptius, Restriction Enzyme
R1632
Mva I from Micrococcus varians Rfl 19, Restriction Enzyme
R8761 Nco I from Nocardia corallina, Restriction Enzyme
R5634
Nhe I from Neisseria mucosa heidelbergensis, Restriction Enzyme
R8506
Not I from Nocardia otidiscaviarum, Restriction Enzyme
R5884 Nsi I from Neisseria sicca, Restriction Enzyme
R7023
Pst I from Providencia stuartii, Restriction Enzyme
R1508
Pvu I from Proteus vulgaris, Restriction Enzyme
R2631
Pvu II from Proteus vulgaris, Restriction Enzyme
R4756 Rsa I from Rhodopseudomonas sphaeroides, Restriction Enzyme
R0754 Sal I from Streptomyces albus G, Restriction Enzyme
R8256
Sfi I from Streptomyces fimbriatus, Restriction Enzyme
R7135 Sph I from Streptomyces phaeochromogenes, Restriction Enzyme
R7260
Xba I from Xanthomonas badrii, Restriction Enzyme
R6379
Xho I from Xanthomonas holcicola, Restriction Enzyme