EMAIL THIS PAGE TO A FRIEND
Stem Cells and Development
Isolation and characterization of novel, highly proliferative human CD34/CD73-double-positive testis-derived stem cells for cell therapy.
PMID 23509942