EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Difficult mask ventilation and muscle paralysis.
PMID 23511535
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8251
Succinylcholine chloride dihydrate, 98.0-102.0%, solid
C14H30Cl2N2O4 · 2H2O