EMAIL THIS PAGE TO A FRIEND
Aviation, space, and environmental medicine
You're the flight surgeon: nasal congestion and Eustachian tube dysfunction.
PMID 23513293
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9652
Terfenadine
C32H41NO2