EMAIL THIS PAGE TO A FRIEND
Journal of the Medical Association of Thailand = Chotmaihet thangphaet
Successful medical treatment in a child with E. coli ESBL meningitis with acute communicating hydrocephalus and ventricular empyema: a case report.
PMID 23513480
Abstract
Extended-spectrum beta-lactamase (ESBL) producing organisms cause wide spectrum of diseases including urinary tract infection, cholangitis, intra-abdominal abscess or pneumonia but rarely meningitis. The present report a successful nonsurgical, medical treatment in a child with Escherichia coli ESBL meningitis with acute symptomatic communicating hydrocephalus and ventricular empyema. Incidence of infections from ESBL producing organisms are increasingly emerging and causing wide spectrum of illnesses which prompts for both aggressive medical and surgical intervention to prevent morbidity and mortality. Antimicrobial agents must be vigilantly utilized to prevent possible development of new highly-resistant organisms.