EMAIL THIS PAGE TO A FRIEND
The Journal of nutrition
Resveratrol inhibits IgE-mediated basophilic mast cell degranulation and passive cutaneous anaphylaxis in mice.
PMID 23514766
Abstract
Resveratrol is a phytoalexin abundantly found in red grape skin and is effective in antitumor and antiinflammation associated with immune responses. This study investigated whether resveratrol suppressed immunoglobulin (Ig)E-mediated allergic responses and passive cutaneous anaphylaxis (PCA) in rat RBL-2H3 mast cells and in BALB/c mice. The release of β-hexosaminidase and histamine was enhanced in mast cells sensitized with anti-dinitrophenyl (DNP)-IgE and subsequently stimulated by DNP-human serum albumin (HSA), indicative of mast cell degranulation. When mast cells were pretreated with nontoxic resveratrol at 1-25 μmol/L, such induction was dose dependently diminished. Spleen tyrosine kinase (Syk) and phospholipase Cγ (PLCγ) of sensitized mast cells were activated by stimulation with DNP-HSA antigen, which was dampened by ≥5 μmol/L resveratrol. The phosphorylation of protein kinase C (PKC)μ and PKCθ was attenuated by administering resveratrol to DNP-HSA-exposed mast cells, whereas quiescent PKCζ/λ in sensitized cells was dose-dependently activated by resveratrol. Male BALB/c mice were sensitized for 24 h with DNP-IgE and orally administered with resveratrol 1 h before the DNP-HSA challenge. The histamine concentration was enhanced in sensitized mice challenged to DNP-HSA, which was reversed by administration of 10 mg/kg resveratrol. Additionally, it encumbered the tissue activation of Syk, PLCγ, and PKCμ in antigen-exposed mice. Resveratrol decreased IgE-mediated PCA and alleviated allergic edema of mouse ear and dorsal skin. Mast cell degranulation and allergic inflammation, accompanying the induction of monocyte chemotactic protein-1 and macrophage inflammatory protein-2, were inhibited by supplementing resveratrol to antigen-challenged mice. Resveratrol inhibited mast cell-derived, immediate-type allergic reactions, and these responses of resveratrol suggest possible therapeutic strategies in preventing allergic inflammatory diseases.