EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Is it the end of the road for synthetic starches in critical illness? No place for hydroxyethyl starch solutions in treatment of patients with sepsis.
PMID 23516156
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6382
Hydroxyethyl starch