EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Divergent reaction pathways of homologous and isosteric propargyl amides in sequential Ru/Pd-catalyzed annulations for the synthesis of heterocycles.
PMID 23521584
Abstract
Cu-catalyzed three-component coupling of imines with benzoyl chloride and terminal arylalkynes followed by enyne ring-closing metathesis (RCM) and Heck cyclization afforded medicinally relevant benzoindolines, cyclopropane-fused indenopyridines, pyrroloquinolines, or 1,7-tetrahydrophenanthrolines via divergent cyclization pathways. Unexpectedly, the Pd-catalyzed cyclization of heterocyclic dienes proceeded via regiodivergent 5-exo or 6-endo pathways depending on the ring size (n = 1, 2) or the presence of isosteric groups (CH vs N). A one-pot protocol for the enyne-RCM/Heck annulation featuring a sequential addition of the Ru and Pd catalysts was developed maximizing the synthetic efficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M74253
N-Methyl-N-propargylbenzylamine, 97%
C11H13N
373192
Palladium, evaporation slug, diam. × L 0.9 cm × 1.2 cm, 99.95% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
348678
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
203998
Palladium, granular, 99.99% trace metals basis
Pd
203939
Palladium, powder, 99.999% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
346985
Palladium, rod, diam. 6.25 mm, ≥99.9% trace metals basis
Pd
267082
Palladium, sponge, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
520810
Palladium black, surface area 40-60 m2/g, 99.95% trace metals basis
Pd
P8013
Pargyline hydrochloride
C11H13N · HCl