EMAIL THIS PAGE TO A FRIEND
Current medicinal chemistry
Chymase inhibition attenuates monocrotaline-induced sinusoidal obstruction syndrome in hamsters.
PMID 23521680
Abstract
Chymase stored in mast cells activates matrix metalloproteinase (MMP)-9, which may relate to the progression of sinusoidal obstruction syndrome (SOS). We investigated the preventive effect of a chymase inhibitor, TY-51469, on monocrotaline-induced SOS in hamsters. Hamsters were orally administrated with a single dose of monocrotaline (120 mg/kg) to induce SOS. Treatment with TY-51469 (1 mg/kg per day) or placebo had started 3 days before the monocrotaline administration. Two days after the monocrotaline administration, significant increases in aspartate aminotransferase, alanine aminotransferase and total bilirubin and a significant reduction of albumin were observed in plasma, but their changes were significantly attenuated by treatment with TY-51469. The numerous hepatic necrosis areas were observed in the placebo-treated group, but the ratio of necrotic area to total area in liver had been significantly reduced by treatment with TY-51469. Both chymase activity and MMP-9 level in liver were significantly augmented in the placebo-treated group. Furthermore, tumor necrosis factor (TNF)-α level in liver was also augmented in the placebo-treated group. However, the chymase activity and levels of MMP-9 and TNF-α were significantly attenuated in the TY-51469-treated group. Until 14 days after monocrotaline administration, survival rates in the placebo- and TY-51469-treated groups were 25% and 70%, respectively, and a significant difference was observed. In conclusion, chymase inhibition by TY-51469 may prevent the accelerating of severity in monocrotaline-induced SOS in hamsters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2401
Crotaline
C16H23NO6
37024
Monocrotaline, analytical standard
C16H23NO6