EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Clinical safety of rapid sequential administration of moxidectin injection and oral derquantel-abamectin as a quarantine treatment for introduced sheep.
PMID 23525484
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31732
Abamectin, PESTANAL®, analytical standard