EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Nanopore-spanning lipid bilayers on silicon nitride membranes that seal and selectively transport ions.
PMID 23528109
Abstract
We report the formation of POPC lipid bilayers that span 130 nm pores in a freestanding silicon nitride film supported on a silicon substrate. These solvent-free lipid membranes self-assemble on organosilane-treated Si3N4 via the fusion of 200 nm unilamellar vesicles. Membrane fluidity is verified by fluorescence recovery after photobleaching (FRAP), and membrane resistance in excess of 1 GΩ is demonstrated using electrical impedance spectroscopy (EIS). An array of 40,000 membranes maintained high impedance over 72 h, followed by rupture of most of the membranes by 82 h. Membrane incorporation of gramicidin, a model ion channel, resulted in increased membrane conductance. This membrane conductance was diminished when the gramicidin channels were blocked with CaCl2, indicating that the change in membrane conductance results from gramicidin-mediated ion transport. These very stable, biologically functional pore-spanning membranes open many possibilities for silicon-based ion-channel devices for applications such as biosensors and high-throughput drug screening.