EMAIL THIS PAGE TO A FRIEND
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova
[Vasobral in the treatment of brain lesions: views of physicians and patients].
PMID 23528488
Abstract
A study was based on the survey of 419 neurologists and 1189 their patients with different forms of cerebrovascular diseases using a specially developed questionnaire. In most cases, vasobral was used as a monotherapy or in a complex treatment. Twenty-two percent of physicians reported that vasobral was the most effective compared to other drugs. The good tolerability of treatment (18%) and the broad spectrum of indications and clinical effects (17%) were reported as well. The large percentage (75%) of patients indicated the positive effect of vasobral on memory, reasoning, vertigo etc The maximal effect was identified in the treatment of mild cognitive impairment caused by chronic brain ischemia and vertebrobasilar insufficiency. Vasobral is recommended for a use in a complex therapy in patients with more severe brain lesions and cognitive deficit.