EMAIL THIS PAGE TO A FRIEND
JAMA internal medicine
Rapid assessment of cardiovascular risk among users of smoking cessation drugs within the US Food and Drug Administration's Mini-Sentinel program.
PMID 23529063