EMAIL THIS PAGE TO A FRIEND
Anaesthesia and intensive care
Management of near fatal mixed tricyclic antidepressant and selective serotonin reuptake inhibitor overdose with Intralipid®20% emulsion.
PMID 23530797