EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Concentration of DNA solutions by extraction with 2-butanol.
PMID 235311
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19440
2-Butanol, puriss. p.a., reag. Ph. Eur., ≥99.5% (GC)
C4H10O