PloS one
A highly selective and non-reaction based chemosensor for the detection of Hg2+ ions using a luminescent iridium(III) complex.
PMID 23533670
Abstract
We report herein a novel luminescent iridium(III) complex with two hydrophobic carbon chains as a non-reaction based chemosensor for the detection of Hg(2+) ions in aqueous solution (<0.002% of organic solvent attributed to the probe solution). Upon the addition of Hg(2+) ions, the emission intensity of the complex was significantly enhanced and this change could be monitored by the naked eye under UV irradiation. The iridium(III) complex shows high specificity for Hg(2+) ions over eighteen other cations. The system is capable of detecting micromolar levels of Hg(2+) ions, which is within the range of many chemical systems.