EMAIL THIS PAGE TO A FRIEND
Ukrains'kyi biokhimichnyi zhurnal (1999 )
Biochemical effects of combined action of gamma-irradiation and paclitaxel on anaplastic thyroid cancer cells.
PMID 23534290
Abstract
The aim of the paper was to describe the biochemical effects of Paclitaxel (Ptx), gamma-irradiation (IR) and their combination in undifferentiated thyroid cancer cells (ATC). IR activated common DNA damage-induced signaling and manifested certain mitogenic effect by inactivation of retinoblastoma protein (pRb). There was clear antagonism between Ptx and IR relative to cell cycle regulators--tumor suppressor p53, pRb, CHK2 and c-Abl as well as proapoptotic Bax expression, but combined action of both agents enhanced caspase-3 and, especially, caspase-8 activation. The Ptx at low (1-25 nM) concentrations caused noticeable radioprotective effect. Thus, in ATC cells the ionizing radiation and Ptx exhibited competitive effects upon phosphorylation of cell cycle controllers: p53, pRb, CHK2, cAbl and expression of Bax. At the same time, the combined effect of radiation and Ptx enhanced antiapoptotic Bcl-2 phosphorylation, caspases activation and survivin expression. The net effect of these events during the first 48-72 h of cells incubation can be considered as antiapoptotic--Ptx attenuated cytotoxic effect of IR.