EMAIL THIS PAGE TO A FRIEND
Biomedical and environmental sciences : BES
Potent in vitro interference of fleroxacin in DNA-binding, unwinding and ATPase activities of Bloom helicase.
PMID 23534463
Abstract
To study the effect of fleroxacin (FLRX) on biological properties of Bloom (BLM) helicase catalytic core (BLM642-1290 helicase) in vitro and the molecular mechanism of interaction between the two molecules. DNA-binding and unwinding activities of BLM642-1290 helicase were assayed by fluorescence polarization and gel retardation assay under conditions that the helicase was subjected to different concentrations of FLRX. Effect of FLRX on helicase ATPase activity was analyzed by phosphorus-free assay based on a colorimetric estimation of ATP hydrolysis-produced inorganic phosphate. Molecular mechanism of interaction between the two molecules was assayed by ultraviolet and fluorescence spectra. The DNA unwinding and ATPase activities of BLM642-1290 helicase were inhibited whereas the DNA-binding activity was promoted in vitro. A BLM-FLRX complex was formed through one binding site, electrostatic and hydrophobic interaction force. Moreover, the intrinsic fluorescence of the helicase was quenched by FLRX as a result of non-radioactive energy transfer. The biological activity of helicase was affected by FLRX, which may be through an allosteric mechanism and stabilization of enzyme conformation in low helicase activity state, disruption of the coupling of ATP hydrolysis to unwinding, and blocking helicase translocation on DNA strands. FLRX may affect the biological activities and conformation of BLM642-1290 helicase, and DNA helicase may be used as a promising drug target for some diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33930
Fleroxacin, VETRANAL, analytical standard
C17H18F3N3O3
F6807
Fleroxacin
C17H18F3N3O3