EMAIL THIS PAGE TO A FRIEND
Biomedical and environmental sciences : BES
Expression of recombinant human lysozyme-tachyplesin I (hLYZ-TP I) in Pichia pastoris and analysis of antibacterial activity.
PMID 23534474