EMAIL THIS PAGE TO A FRIEND
Clinical nephrology
Pseudo-blood leak? A hemodialysis mystery.
PMID 23537683
Abstract
Hydroxocobalamin is a treatment for cyanide toxicity with few side effects. We report a case of a hemodialysis patient whose treatment was compromised by hydroxocobalamin interference with the blood leak detector.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1428000 Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
C62H89CoN13O15P