EMAIL THIS PAGE TO A FRIEND
Gastroenterologia y hepatologia
[Split-dose sodium picosulphate/magnesium citrate for morning colonoscopies performed 2 to 6 hours after fluid intake].
PMID 23537750
Abstract
Split dosage of bowel preparations has been shown to substantially improve bowel cleansing. To compare the split dose (SD) sodium picosulphate/magnesium oxide/anhydrous citric acid (Citrafleet(®)) regimen for morning colonoscopies with standard cleansing the day before. Consecutive outpatients were randomized to receive Citrafleet(®) the day before colonoscopy or SD, in whom the second half was administered on an individual basis from 2 to 6 hours before the procedure. No bisacodyl was administered. All procedures were performed with non-anesthesiologist administered propofol sedation. The Boston scale was used to assess the quality of bowel preparation (adequate cleansing if score ≥ 6, with no score of 0/1 in any segment). A total of 193 patients were included. Overall bowel cleansing was significantly better in the SD group (7 vs. 5.2, p<0.001), as well as in the cecum (2.4 vs. 1.4, p < 0.001), ascending colon (2.5 vs. 1.6, p<0.001) and transverse colon (2.4 vs. 2, p=0.004). A significant proportion of SD patients had adequate bowel cleansing (71% vs. 30%, p<0.001). Patients in the SD group drank a greater amount of liquid (4.9 vs. 4 liters, p=0.006) and more frequently perceived the cleansing process to be easy or very easy to complete (89 vs. 68%, p=0.04), although they slept significantly fewer hours (6.5 vs. 7.9, p<0.001). No bronchoaspiration pneumonia was reported. SD Citrafleet(®) 2 to 6 hours before colonoscopy increased the rate of procedures with adequate bowel cleansing by 40%, especially in the proximal colon, allowed more liquids to be drunk and increased the perception of ease in completing the preparation, with no sedation-related complications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001104 Picosulfate for system suitability, European Pharmacopoeia (EP) Reference Standard
C18H13NNa2O8S2