EMAIL THIS PAGE TO A FRIEND
Genome announcements
Draft Whole-Genome Sequence of Bacillus sonorensis Strain L12, a Source of Nonribosomal Lipopeptides.
PMID 23538904
Abstract
The Bacillus sonorensis L12 draft genome sequence is approximately 4,647,754xa0bp in size with a G+C content of 45.2%. Over 86% of the genome contains protein-encoding genes, including several gene clusters for de novo biosynthesis of the nonribosomal lipopeptides iturin, bacitracin, and fengycin, which could mean that the strain exhibits antifungal effects.
Related Materials