EMAIL THIS PAGE TO A FRIEND
Indian journal of pediatrics
Use of continuous hemodiafiltration for the treatment of a neonate with hyperammonemia secondary to argininosuccinate lyase (ASAL) deficiency.
PMID 23539248