EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Roots and associated fungi drive long-term carbon sequestration in boreal forest.
PMID 23539604
Abstract
Boreal forest soils function as a terrestrial net sink in the global carbon cycle. The prevailing dogma has focused on aboveground plant litter as a principal source of soil organic matter. Using (14)C bomb-carbon modeling, we show that 50 to 70% of stored carbon in a chronosequence of boreal forested islands derives from roots and root-associated microorganisms. Fungal biomarkers indicate impaired degradation and preservation of fungal residues in late successional forests. Furthermore, 454 pyrosequencing of molecular barcodes, in conjunction with stable isotope analyses, highlights root-associated fungi as important regulators of ecosystem carbon dynamics. Our results suggest an alternative mechanism for the accumulation of organic matter in boreal forests during succession in the long-term absence of disturbance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47130-U
Ergosterol, 10 mg/mL in chloroform, analytical standard
C28H44O