EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Busting myths about UK food production.
PMID 23539646
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8648 Somatotropin (STH) from porcine pituitary