EMAIL THIS PAGE TO A FRIEND
Canadian journal of microbiology
Biofilms from a Brazilian water distribution system include filamentous fungi.
PMID 23540336
Abstract
Filamentous fungi in drinking water can block water pipes, can cause organoleptic biodeterioration, and are a source of pathogens. There are increasing reports of the involvement of the organisms in biofilms. This present study describes a sampling device that can be inserted directly into pipes within water distribution systems, allowing biofilm formation in situ. Calcofluor White M2R staining and fluorescent in situ hybridization with morphological analyses using epifluorescent microscopy were used to analyse biofilms for filamentous fungi, permitting direct observation of the fungi. DAPI (4',6-diamidino-2-phenylindole) was applied to detect bacteria. Filamentous fungi were detected in biofilms after 6 months on coupons exposed to raw water, decanted water and at the entrance of the water distribution system. Algae, yeast, and bacteria were also observed. The role of filamentous fungi requires further investigations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3543
Fluorescent Brightener 28, used as a stain and brightening agent
C40H44N12O10S2