EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Evaluation of tropicamide-loaded tamarind seed xyloglucan nanoaggregates for ophthalmic delivery.
PMID 23544540
Abstract
The present study was aimed to prepare tamarind seed nanoaggregates and its evaluation for ophthalmic delivery. The preparation of tropicamide-loaded tamarind seed xyloglucan nanoaggregates was optimized using face centred central composite experimental design, employing the concentrations of tamarind seed xyloglucan and Poloxamer-407, as independent variables. The results revealed that concentration of TSX has a significant antagonistic effect on particle size, while poloxamer displayed a significant synergistic effect on encapsulation efficiency. The optimal concentrations of TSX and poloxamer were found to be 0.45% (w/v) and 0.5% (w/v) respectively. The optimized formulation of tropicamide-loaded TSX nanoaggregates showed a significantly higher corneal permeation of tropicamide across the isolated goat cornea compared to commercial conventional aqueous formulation. The results revealed excellent mucoadhesive properties of TSX nanoaggregates. Further, the tropicamide-loaded TSX nanoaggregates formulation showed excellent ocular tolerance and biocompatibility as determined by hen's egg test chorioallantoic membrane and resazurin assay on Vero cell lines.