EMAIL THIS PAGE TO A FRIEND
Neurosciences (Riyadh, Saudi Arabia)
Diagnostic uncertainty of tumefactive cystic demyelinating lesions.
PMID 23545620
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1755000 Methylprednisolone acetate
C24H32O6