EMAIL THIS PAGE TO A FRIEND
Optics express
Enhanced performance of InGaN/GaN based solar cells with an In(0.05)Ga(0.95)N ultra-thin inserting layer between GaN barrier and In(0.2)Ga(0.8)N well.
PMID 23546093
Abstract
The effect of ultra-thin inserting layer (UIL) on the photovoltaic performances of InGaN/GaN solar cells is investigated. With UIL implemented, the open-circuit voltage was increased from 1.4 V to 1.7 V, short-circuit current density was increased by 65% and external quantum efficiency was increased by 59%, compared to its counterparts at room temperature under 1-sun AM1.5G illumination. The improvements in electrical and photovoltaic properties are mainly attributed to the UIL which can boost the crystal quality and alleviate strain. Moreover, it can act as a transition layer for higher indium incorporation and an effective light sub-absorption layer in multiple quantum wells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57083
Indium, bars, 99.95%
In
264113
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
In
326607
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
In
357278
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
In
326631
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
In
357286
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
In
264040
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
In
357294
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
In
264059
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
In
326615
Indium, pieces, 99.99% trace metals basis
In
203432
Indium, powder, −60 mesh, 99.999% trace metals basis
In
264032
Indium, powder, −100 mesh, 99.99% trace metals basis
In
277959
Indium, powder, 99.99% trace metals basis
In
385379
Indium, rod, diam. 10 mm, 99.9998% trace metals basis
In
264091
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
In
357065
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
In
340863
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
In
357073
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
In
264075
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
In
264067
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
In
326623
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
In