EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthetic route to chiral indolines via ring-opening/C-N cyclization of activated 2-haloarylaziridines.
PMID 23548056
Abstract
A practical approach for the synthesis of 3-substituted indolines via regio- and stereoselective SN2-type ring-opening of 2-(2-halophenyl)-N-tosylaziridines with heteroatomic nucleophiles (O, N, and S) followed by palladium-catalyzed intramolecular C-N cyclization is reported in excellent yields (up to >99%) and enantiomeric excess (ee 99%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5605
Indoline, ReagentPlus®, 99%
C8H9N