EMAIL THIS PAGE TO A FRIEND
Cancer letters
Guggulsterone sensitizes glioblastoma cells to Sonic hedgehog inhibitor SANT-1 induced apoptosis in a Ras/NFκB dependent manner.
PMID 23548480
Abstract
Since Shh pathway effector, Gli1, is overexpressed in gliomas, we investigated the effect of novel Shh inhibitor SANT-1 on glioma cell viability. Though SANT-1 failed to induce apoptosis, it reduced proliferation of glioma stem-like cells. Apart from canonical Shh cascade, Gli1 is also induced by non-canonical pathways including NFκB. Therefore, a combinatorial strategy with Ras/NFκB inhibitor, Guggulsterone, was employed to enhance effectiveness of SANT-1. Guggulsterone inhibited Ras and NFκB activity and sensitized cells to SANT-1 induced apoptosis via intrinsic apoptotic mechanism. Inhibition of either Ras or NFκB activity was sufficient to sensitize cells to SANT-1. Guggulsterone induced ERK activation also contributed to Caspase-9 activation. Since SANT-1 and Guggulsterone differentially target stem-like and non-stem glioma cells respectively, this combination warrants investigation as an effective anti-glioma therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4572
SANT-1, ≥98% (HPLC), powder
C23H27N5