EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Goji berry: a potential new player in latex-food syndrome.
PMID 23548535