EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Autocrine production of IL-11 mediates tumorigenicity in hypoxic cancer cells.
PMID 23549086
Abstract
IL-11 and its receptor, IL-11Ra, are expressed in human cancers; however, the functional role of IL-11 in tumor progression is not known. We found that IL11 is a hypoxia-inducible, VHL-regulated gene in human cancer cells and that expression of IL11 mRNA was dependent, at least in part, on HIF-1. A cooperative interaction between HIF-1 and AP-1 mediated transcriptional activation of the IL11 promoter. Additionally, we found that human cancer cells expressed a functional IL-11Ra subunit, which triggered signal transduction either by exogenous recombinant human IL-11 or by autocrine production of IL-11 in cells cultured under hypoxic conditions. Silencing of IL11 dramatically abrogated the ability of hypoxia to increase anchorage-independent growth and significantly reduced tumor growth in xenograft models. Notably, these results were phenocopied by partial knockdown of STAT1 in a human prostate cancer cell line (PC3), suggesting that this pathway may play an important role in mediating the effects of IL-11 under hypoxic conditions. In conclusion, these results identify IL11 as an oxygen- and VHL-regulated gene and provide evidence of a pathway "hijacked" by hypoxic cancer cells that may contribute to tumor progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3934 Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
C7025 Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
C2273 Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
C5024 Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
C2624 Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
C4396 Carbonic Anhydrase I from human erythrocytes
C6653 Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
C7749 Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
C6624 Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution
C3640 Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
C3666 Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
C2522 Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
C6165 Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
CKOZFN10563 CompoZr® Knockout ZFN Kit, Human IL11 (NM_000641)